6-Methyl-2-thiouracil ≥95%

Supplier: Matrix Scientific

Synonyms: Methiocil, 2-Mercapto-6-methylpyrimidin-4(1H)-one, Methylthiouracil, MZU, Basethyrin, 4-Hydroxy-2-mercapto 6-methylpyrimidine, Thiothymin

073074-1G 073074-5G 073074-10G
102528-260EA 55.38 USD
102528-260 102528-262 102528-264
6-Methyl-2-thiouracil ≥95%
6-Methyl-2-thiouracil

Hazard: Irritant

Formula: C₅H₆N₂OS
MW: 142.18 g/mol
MDL Number: MFCD00006040
CAS Number: 56-04-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR