2-Bromo-6-nitrotoluene ≥95%

Supplier: Matrix Scientific

073073-100G 073073-25G
102528-258EA 526.15 USD
102528-258 102833-588
2-Bromo-6-nitrotoluene ≥95%
2-Bromo-6-nitrotoluene

Hazard: Irritant

Formula: C₇H₆BrNO₂
MW: 216.03 g/mol
Boiling Pt: 142-147°C/30 Torr
Melting Pt: 37-42°C
MDL Number: MFCD00009792
CAS Number: 55289-35-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR