6-Fluoro-o-toluidine ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Fluoro-6-methylaniline

073072-5G 073072-25G 073072-100G
102528-250EA 30.77 USD
102528-250 102663-922 102663-924
6-Fluoro-o-toluidine ≥95%
6-Fluoro-o-toluidine

Hazard: Irritant

Formula: C₇H₈FN
MW: 125.1435 g/mol
Density: 1.082
Flash Pt: 76°C
MDL Number: MFCD06658252
CAS Number: 443-89-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR