6-Fluoro-3,4-dihydro-2H-benzo[1,4]oxazine ≥95%

Supplier: Matrix Scientific

073069-250MG 073069-1G 073069-5G
102528-232EA 192.31 USD
102528-232 102528-234 102528-236
6-Fluoro-3,4-dihydro-2H-benzo[1,4]oxazine ≥95%
6-Fluoro-3,4-dihydro-2H-benzo[1,4]oxazine

Hazard: Irritant

Formula: C₈H₈FNO
CAS Number: 105655-00-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR