4-Chloro-2,6-dimethylpyridine ≥95%

Supplier: Matrix Scientific

073064-1G 073064-5G
102528-204EA 92.31 USD
102528-204 102528-206
4-Chloro-2,6-dimethylpyridine ≥95%
4-Chloro-2,6-dimethylpyridine

Hazard: Irritant

Formula: C₇H₈ClN
MDL Number: MFCD01646174
CAS Number: 3512-75-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR