6-Bromo-8-chloroimidazo[1,2-a]pyrazine ≥95%

Supplier: Matrix Scientific

073063-250MG 073063-1G
102528-200EA 967.69 USD
102528-200 102528-202
6-Bromo-8-chloroimidazo[1,2-a]pyrazine ≥95%
6-Bromo-8-chloroimidazo[1,2-a]pyrazine

Hazard: Irritant

Formula: C₆H₃BrClN₃
MW: 232.4700 g/mol
MDL Number: MFCD11846485
CAS Number: 1208083-37-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR