N,N'-Dimethylethylendiamine ≥95%

Supplier: Matrix Scientific

Synonyms: N1,N2-Dimethylethane-1,2-diamine, sym. Dimethylethylendiamine, N,N'-Dimethylethylenediamine

073060-100G 073060-25G
102528-190EA 176.92 USD
102528-190 102663-918
N,N'-Dimethylethylendiamine ≥95%
N,N'-Dimethylethylendiamine

Hazard: Flammable/Corrosive/Air Sensitive/Stench

Formula: C₄H₁₂N₂
MW: 88.15 g/mol
Boiling Pt: 118-119°C
Density: 0.818
Flash Pt: 28°C
MDL Number: MFCD00008290
CAS Number: 110-70-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR