3-Amino-6-chloro-2-pyrazinecarbonitrile ≥95%

Supplier: Matrix Scientific

073059-250MG 073059-1G 073059-5G
102528-182EA 461.54 USD
102528-182 102528-184 102528-186
3-Amino-6-chloro-2-pyrazinecarbonitrile ≥95%
3-Amino-6-chloro-2-pyrazinecarbonitrile

Hazard: Irritant

Formula: C5H3ClN4
MW: 154.5600 g/mol
MDL Number: MFCD10697784
CAS Number: 17231-50-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR