2-Chloro-7-methylquinoxaline ≥95%
Supplier: Matrix Scientific
073058-250MG
073058-1G
073058-5G
102528-176EA
456.92
USD
102528-176
102528-178
102528-180
2-Chloro-7-methylquinoxaline ≥95%
2-Chloro-7-methylquinoxaline
Hazard: Irritant
Formula:
C₉H₇ClN₂ |
CAS Number:
90272-84-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...