2,8-Dichloroquinoxaline ≥95%

Supplier: Matrix Scientific

073057-250MG 073057-1G
102528-172EA 826.15 USD
102528-172 102528-174
2,8-Dichloroquinoxaline ≥95%
2,8-Dichloroquinoxaline

Hazard: Irritant

Formula: C₈H₄Cl₂N₂
MDL Number: MFCD11846471
CAS Number: 120258-69-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR