2-Chloro-6-nitroquinoxaline ≥95%

Supplier: Matrix Scientific

073056-250MG 073056-1G 073056-5G
102528-166EA 461.54 USD
102528-166 102528-168 102528-170
2-Chloro-6-nitroquinoxaline ≥95%
2-Chloro-6-nitroquinoxaline

Hazard: Irritant

Formula: C₈H₄ClN₃O₂
MW: 209.5900 g/mol
MDL Number: MFCD03067026
CAS Number: 6272-25-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR