(1R,4S)-2-Azabicyclo[2.2.1]hept-5-en-3-one ≥95%

Supplier: Matrix Scientific

Synonyms: (-)-2-Azabicyclo[2.2.1]hept-5-en-3-one

073054-1G 073054-5G 073054-10G
102528-154EA 52.31 USD
102528-154 102528-156 102528-158
(1R,4S)-2-Azabicyclo[2.2.1]hept-5-en-3-one ≥95%
(1R,4S)-2-Azabicyclo[2.2.1]hept-5-en-3-one

Hazard: Irritant

Formula: C₆H₇NO
Melting Pt: 94…95 °C
MDL Number: MFCD00211274
CAS Number: 79200-56-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR