5'-Deoxy-5-fluorocytidine ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Amino-1-((2R,3R,4S,5R)-3,4-dihydroxy-5-methyltetrahydrofuran-2-yl)-5-fluoropyrimidin-2(1H)-one

073053-1G 073053-5G 073053-10G
102528-148EA 213.85 USD
102528-148 102528-150 102528-152
5'-Deoxy-5-fluorocytidine ≥95%
5'-Deoxy-5-fluorocytidine

Hazard: Irritant

Formula: C₉H₁₂FN₃O₄
MW: 245.21 g/mol
MDL Number: MFCD07369278
CAS Number: 66335-38-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR