6-Bromochromen-2-one ≥95%

Supplier: Matrix Scientific

073051-1G 073051-5G
102528-140EA 710.77 USD
102528-140 102528-142
6-Bromochromen-2-one ≥95%
6-Bromochromen-2-one

Hazard: Irritant

Formula: C₉H₅BrO₂
MW: 225.0388 g/mol
MDL Number: MFCD01327473
CAS Number: 19063-55-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR