N-Methyl-1-(pyrimidin-2-yl)methanamine ≥95%

Supplier: Matrix Scientific

073046-250MG 073046-1G 073046-5G
102528-112EA 640 USD
102528-112 102528-114 102528-116
N-Methyl-1-(pyrimidin-2-yl)methanamine ≥95%
N-Methyl-1-(pyrimidin-2-yl)methanamine

Hazard: Irritant

Formula: C₆H₉N₃
MW: 123.1600 g/mol
MDL Number: MFCD11617315
CAS Number: 1083246-53-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR