Benzo[d]isothiazol-3-amine ≥95%

Supplier: Matrix Scientific

073045-250MG 073045-1G 073045-5G
102528-106EA 1383.08 USD
102528-106 102528-108 102528-110
Benzo[d]isothiazol-3-amine ≥95%
Benzo[d]isothiazol-3-amine

Hazard: Irritant

Formula: C₇H₆N₂S
MW: 150.2009 g/mol
MDL Number: MFCD00726295
CAS Number: 23031-78-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR