4-(3-Bromoimidazo[1,2-b]pyridazin-6-yl)morpholine ≥95%

Supplier: Matrix Scientific

073044-250MG 073044-1G 073044-5G
102528-100EA 1535.38 USD
102528-100 102528-102 102528-104
4-(3-Bromoimidazo[1,2-b]pyridazin-6-yl)morpholine ≥95%
4-(3-Bromoimidazo[1,2-b]pyridazin-6-yl)morpholine

Hazard: Irritant

Formula: C₁₀H₁₁BrN₄O
MW: 283.1300 g/mol
MDL Number: MFCD11520886
CAS Number: 1012343-72-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR