3-Bromoimidazo[1,2-b]pyridazin-6-ylamine ≥95%

Supplier: Matrix Scientific

073043-250MG 073043-1G 073043-5G
102528-094EA 198.46 USD
102528-094 102528-096 102528-098
3-Bromoimidazo[1,2-b]pyridazin-6-ylamine ≥95%
3-Bromoimidazo[1,2-b]pyridazin-6-ylamine

Hazard: Irritant

Formula: C6H5BrN4
MW: 213.0400 g/mol
MDL Number: MFCD11520885
CAS Number: 1260850-70-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR