5-Bromo-2-methyl-2H-indazole-3-carbaldehyde ≥95%

Supplier: Matrix Scientific

073042-250MG 073042-1G 073042-5G
102528-088EA 967.69 USD
102528-088 102528-090 102528-092
5-Bromo-2-methyl-2H-indazole-3-carbaldehyde ≥95%
5-Bromo-2-methyl-2H-indazole-3-carbaldehyde

Hazard: Irritant

Formula: C₉H₇BrN₂O
MW: 239.0700 g/mol
MDL Number: MFCD11520882
CAS Number: 1251023-52-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR