6-Bromo-2H-1,4-benzoxazin-3(4H)-one ≥95%

Supplier: Matrix Scientific

073040-1G 073040-5G 073040-10G
102528-076EA 143.08 USD
102528-076 102528-078 102528-080
6-Bromo-2H-1,4-benzoxazin-3(4H)-one ≥95%
6-Bromo-2H-1,4-benzoxazin-3(4H)-one

Hazard: Irritant

Formula: C₈H₆BrNO₂
MDL Number: MFCD00461173
CAS Number: 24036-52-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR