6-Bromo-4H-3,1-benzoxazin-4-one ≥95%

Supplier: Matrix Scientific

073038-250MG 073038-1G 073038-5G
102528-064EA 355.38 USD
102528-064 102528-066 102528-068
6-Bromo-4H-3,1-benzoxazin-4-one ≥95%
6-Bromo-4H-3,1-benzoxazin-4-one

Hazard: Irritant

Formula: C₈H₄BrNO₂
CAS Number: 449185-77-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR