2-(Methylthio)pyrimidine-5-carboxylic acid ≥95%

Supplier: Matrix Scientific

073037-1G 073037-5G
102528-060EA 604.62 USD
102528-060 102528-062
2-(Methylthio)pyrimidine-5-carboxylic acid ≥95%
2-(Methylthio)pyrimidine-5-carboxylic acid

Hazard: Irritant

Formula: C₆H₆N₂O₂S
MDL Number: MFCD07394096
CAS Number: 110099-94-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR