2-Amino-5-fluoro-3-methylpyridine ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Fluoro-3-methylpyridin-2-ylamine

073036-250MG 073036-1G 073036-5G
102528-054EA 183.08 USD
102528-054 102528-056 102528-058
2-Amino-5-fluoro-3-methylpyridine ≥95%
2-Amino-5-fluoro-3-methylpyridine

Hazard: Irritant

Formula: C₆H₇FN₂
MW: 126.1300 g/mol
MDL Number: MFCD07375130
CAS Number: 886365-56-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR