2H-1,2,4-Triazole-3-carbonitrile ≥95%

Supplier: Matrix Scientific

Synonyms: 2H-1,2,4-triazole-3-carbonitrile

073030-1G 073030-5G 073030-10G
102528-020EA 46.15 USD
102528-020 102528-022 102528-024
2H-1,2,4-Triazole-3-carbonitrile ≥95%
2H-1,2,4-Triazole-3-carbonitrile

Hazard: Irritant

Formula: C₃H₂N₄
MW: 94.0700 g/mol
Melting Pt: 187 °C
MDL Number: MFCD11505654
CAS Number: 3641-10-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR