2,3-Dibromopyrazine ≥95%

Supplier: Matrix Scientific

073029-250MG 073029-1G 073029-5G
102528-014EA 498.46 USD
102528-014 102528-016 102528-018
2,3-Dibromopyrazine ≥95%
2,3-Dibromopyrazine

Hazard: Irritant

Formula: C₄H₂Br₂N₂
MDL Number: MFCD02684897
CAS Number: 95538-03-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR