Imidazo[1,2-a]pyrazine ≥95%
Supplier: Matrix Scientific
073028-1G
073028-5G
102528-008EA
130.77
USD
102528-008
102528-010
Imidazo[1,2-a]pyrazine ≥95%
Imidazo[1,2-a]pyrazine
Hazard: Irritant
Formula:
C₆H₅N₃ MW: 119.1200 g/mol |
MDL Number:
MFCD06245371 CAS Number: 274-79-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...