6-Chloro-5-nitroquinoline ≥95%

Supplier: Matrix Scientific

073026-250MG 073026-1G 073026-5G
102527-996EA 355.38 USD
102527-996 102527-998 102528-000
6-Chloro-5-nitroquinoline ≥95%
6-Chloro-5-nitroquinoline

Hazard: Irritant

Formula: C₉H₅ClN₂O₂
MW: 208.6012 g/mol
MDL Number: MFCD00093616
CAS Number: 86984-32-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR