5-Bromobenzo[b]thiophene ≥95%

Supplier: Matrix Scientific

073024-1G 073024-5G 073024-10G
102527-984EA 46.15 USD
102527-984 102527-986 102527-988
5-Bromobenzo[b]thiophene ≥95%
5-Bromobenzo[b]thiophene

Hazard: Irritant/Light Sensitive/Refrigerate

Formula: C₈H₅BrS
Boiling Pt: 105…107 °C (10 mmHg)
Melting Pt: 44…48 °C
MDL Number: MFCD03069318
CAS Number: 4923-87-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR