2-Hydroxy-4-methoxybenzonitrile ≥95%

Supplier: Matrix Scientific

073023-1G 073023-5G 073023-10G
102527-978EA 533.85 USD
102527-978 102527-980 102527-982
2-Hydroxy-4-methoxybenzonitrile ≥95%
2-Hydroxy-4-methoxybenzonitrile

Hazard: Irritant

Formula: C₈H₇NO₂
MW: 149.1500 g/mol
MDL Number: MFCD08567959
CAS Number: 39835-11-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR