3-Amino-5-hydroxy-1H-pyrazole ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Amino-5-hydroxy-1H-pyrazole, 3-Amino-5-hydroxypyrazole, 5-Amino-3-pyrazolol, 3-Amino-1H-pyrazol-5(4H)-one, 3-Amino-2-pyrazolin-5-one, 5-Hydroxy-3-aminopyrazole, 5-Amino-2,4-dihydro-3H-pyrazol-3-one

073022-5G 073022-10G 073022-100G
102527-972EA 64.62 USD
102527-972 102527-974 102527-976
3-Amino-5-hydroxy-1H-pyrazole ≥95%
3-Amino-5-hydroxy-1H-pyrazole

Hazard: Irritant

Formula: C₃H₅N₃O
MW: 99.09 g/mol
MDL Number: MFCD00022384
CAS Number: 6126-22-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR