6-Methyl-4-phenylchroman-2-one ≥95%

Supplier: Matrix Scientific

Synonyms: 6-Methyl-4-Phenylchromanone, 3,4-Dihydro-6-methyl-4-phenylcoumarin, 6-Methyl-4-phenyl-2-chromanone

073021-250MG 073021-1G 073021-5G
102527-966EA 355.38 USD
102527-966 102527-968 102527-970
6-Methyl-4-phenylchroman-2-one ≥95%
6-Methyl-4-phenylchroman-2-one

Hazard: Irritant

Formula: C₁₆H₁₄O₂
MW: 238.2812 g/mol
MDL Number: MFCD03695019
CAS Number: 40546-94-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR