2-Methyl-1H-benzoimidazol-5-ylamine ≥95%

Supplier: Matrix Scientific

073013-1G 073013-5G
102527-922EA 710.77 USD
102527-922 102527-924
2-Methyl-1H-benzoimidazol-5-ylamine ≥95%
2-Methyl-1H-benzoimidazol-5-ylamine

Hazard: Irritant

Formula: C₈H₉N₃
MW: 147.1800 g/mol
MDL Number: MFCD00462859
CAS Number: 29043-48-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR