Methyl-3-amino-6-chloropyrazine-2-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 3-amino-6-chloropyrazine-2-carboxylate

073012-1G 073012-5G
102527-918EA 116.92 USD
102527-918 102527-920
Methyl-3-amino-6-chloropyrazine-2-carboxylate ≥95%
Methyl-3-amino-6-chloropyrazine-2-carboxylate

Hazard: Irritant

Formula: C₆H₆ClN₃O₂
MW: 187.58 g/mol
MDL Number: MFCD00834965
CAS Number: 1458-03-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR