7-Fluoro-6-nitroquinazolin-4(3H)-one ≥95%

Supplier: Matrix Scientific

Synonyms: 7-Fluoro-6-nitroquinazolin-4(1H)-one, 7-fluoro-6-nitro-3,4-dihydroquinazolin-4-one, 7-Fluoro-6-nitroquinazolin-4(3H)-one, 7-Fluoro-4-hydroxy-6-nitroquinazoline

073011-1G 073011-5G
102527-912EA 460 USD
102527-912 102527-914
7-Fluoro-6-nitroquinazolin-4(3H)-one ≥95%
7-Fluoro-6-nitroquinazolin-4(3H)-one

Hazard: Irritant

Formula: C₈H₄FN₃O₃
MW: 209.1300 g/mol
MDL Number: MFCD08690890
CAS Number: 162012-69-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR