7-Bromo-1-methyl-1H-quinoxalin-2-one ≥95%

Supplier: Matrix Scientific

073009-250MG 073009-1G
102527-904EA 640 USD
102527-904 102527-906
7-Bromo-1-methyl-1H-quinoxalin-2-one ≥95%
7-Bromo-1-methyl-1H-quinoxalin-2-one

Hazard: Irritant

Formula: C₉H₇BrN₂O
MW: 239.0687 g/mol
MDL Number: MFCD10000855
CAS Number: 82019-32-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR