3-(Aminomethyl)-1H-indazole ≥95%

Supplier: Matrix Scientific

073008-250MG 073008-1G 073008-5G
102527-898EA 427.69 USD
102527-898 102527-900 102527-902
3-(Aminomethyl)-1H-indazole ≥95%
3-(Aminomethyl)-1H-indazole

Hazard: Irritant

Formula: C8H9N3
MW: 147.1772 g/mol
MDL Number: MFCD06738683
CAS Number: 806640-37-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR