4-Chloro-6-methoxyquinazoline ≥95%

Supplier: Matrix Scientific

073007-1G 073007-5G
102527-894EA 710.77 USD
102527-894 102527-896
4-Chloro-6-methoxyquinazoline ≥95%
4-Chloro-6-methoxyquinazoline

Hazard: Irritant

Formula: C₉H₇ClN₂O
MW: 194.6177 g/mol
MDL Number: MFCD06657615
CAS Number: 50424-28-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR