4-Aminoquinoline-2-one ≥95%

Supplier: Matrix Scientific

073006-250MG 073006-1G 073006-5G
102527-888EA 569.23 USD
102527-888 102527-890 102527-892
4-Aminoquinoline-2-one ≥95%
4-Aminoquinoline-2-one

Hazard: Irritant

Formula: C₉H₈N₂O
MDL Number: MFCD05665662
CAS Number: 110216-87-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR