2-(N-Boc-N-methylamino)-5-bromopyridine ≥95%
Supplier: Matrix Scientific
Synonyms:
tert-Butyl (5-bromopyridin-2-yl)(methyl)carbamate
073005-250MG
073005-1G
073005-5G
102527-882EA
64.62
USD
102527-882
102527-884
102527-886
2-(N-Boc-N-methylamino)-5-bromopyridine ≥95%
2-(N-Boc-N-methylamino)-5-bromopyridine
Hazard: Irritant
Formula:
C₁₁H₁₅BrN₂O₂ |
MDL Number:
MFCD12546613 CAS Number: 227939-01-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...