2-Chloro-4-morpholinothieno[3,2-d]pyrimidine-6-carbaldehyde ≥95%

Supplier: Matrix Scientific

073004-250MG 073004-1G 073004-5G
102527-876EA 783.08 USD
102527-876 102527-878 102527-880
2-Chloro-4-morpholinothieno[3,2-d]pyrimidine-6-carbaldehyde ≥95%
2-Chloro-4-morpholinothieno[3,2-d]pyrimidine-6-carbaldehyde

Hazard: Irritant

Formula: C₁₁H₁₀ClN₃O₂S
CAS Number: 885618-31-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR