Quinoxalin-5-ol ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Hydroxyquinoxaline

073001-1G 073001-5G
102527-862EA 710.77 USD
102527-862 102527-864
Quinoxalin-5-ol ≥95%
Quinoxalin-5-ol

Hazard: Irritant

Formula: C8H6N2O
MW: 146.05 g/mol
MDL Number: MFCD00972004
CAS Number: 17056-99-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR