7-Bromoquinoxalin-2(1H)-one ≥95%

Supplier: Matrix Scientific

072999-1G 072999-5G
102527-852EA 238.46 USD
102527-852 102527-854
7-Bromoquinoxalin-2(1H)-one ≥95%
7-Bromoquinoxalin-2(1H)-one

Hazard: Irritant

Formula: C₈H₅BrN₂O
MDL Number: MFCD01465145
CAS Number: 82031-32-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR