5-(4-Methylpiperazin-1-yl)pyridin-2-amine ≥95%

Supplier: Matrix Scientific

Synonyms: 1-Methyl-4-(6-aminopyridin-3-yl)piperazine

072998-1G 072998-5G
102527-846EA 67.69 USD
102527-846 102527-848
5-(4-Methylpiperazin-1-yl)pyridin-2-amine ≥95%
5-(4-Methylpiperazin-1-yl)pyridin-2-amine

Hazard: Irritant/Refrigerate

Formula: C₁₀H₁₆N₄
MW: 192.2608 g/mol
MDL Number: MFCD10009563
CAS Number: 571189-49-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR