4-Methylthiazole-2-carboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Methyl-1,3-thiazole-2-carboxylic acid

072994-1G 072994-5G 072994-10G
102527-822EA 123.08 USD
102527-822 102527-824 102527-826
4-Methylthiazole-2-carboxylic acid ≥95%
4-Methylthiazole-2-carboxylic acid

Hazard: Irritant

Formula: C₅H₅NO₂S
MW: 143.16 g/mol
MDL Number: MFCD07377486
CAS Number: 14542-16-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR