5-Methylthiazole-2-carboxylic acid ≥95%

Supplier: Matrix Scientific

072993-1G 072993-5G
102527-818EA 35.38 USD
102527-818 102527-820
5-Methylthiazole-2-carboxylic acid ≥95%
5-Methylthiazole-2-carboxylic acid

Hazard: Irritant/Store in Freezer

Formula: C5H5NO2S
MW: 143.16 g/mol
MDL Number: MFCD08060736
CAS Number: 61291-21-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR