2-Amino-6-methylpyrazine ≥95%

Supplier: Matrix Scientific

Synonyms: 6-Methylpyrazin-2-amine

072992-1G 072992-5G
102527-814EA 541.54 USD
102527-814 102527-816
2-Amino-6-methylpyrazine ≥95%
2-Amino-6-methylpyrazine

Hazard: Irritant

Formula: C₅H₇N₃
MW: 109.13 g/mol
MDL Number: MFCD08235193
CAS Number: 5521-56-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR