2-(Trifluoromethyl)-5,6,7,8-tetrahydro[1,2,4]triazolo[1,5-a]pyrazine ≥95%

Supplier: Matrix Scientific

072990-250MG 072990-1G
102527-804EA 967.69 USD
102527-804 102527-806
2-(Trifluoromethyl)-5,6,7,8-tetrahydro[1,2,4]triazolo[1,5-a]pyrazine ≥95%
2-(Trifluoromethyl)-5,6,7,8-tetrahydro[1,2,4]triazolo[1,5-a]pyrazine

Hazard: Irritant

Formula: C₆H₇F₃N₄
MW: 192.1400 g/mol
MDL Number: MFCD09834979
CAS Number: 681249-57-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR