4-Chloro-6,7-bis(2-methoxyethoxy)quinazoline ≥95%

Supplier: Matrix Scientific

072989-1G 072989-5G
102527-800EA 69.23 USD
102527-800 102527-802
4-Chloro-6,7-bis(2-methoxyethoxy)quinazoline ≥95%
4-Chloro-6,7-bis(2-methoxyethoxy)quinazoline

Hazard: Irritant

Formula: C₁₄H₁₇ClN₂O₄
MDL Number: MFCD09751265
CAS Number: 183322-18-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR