3-Chloro-2-nitropyridine ≥95%

Supplier: Matrix Scientific

072985-1G 072985-5G
102527-782EA 144.62 USD
102527-782 102527-784
3-Chloro-2-nitropyridine ≥95%
3-Chloro-2-nitropyridine

Hazard: Irritant

Formula: C₅H₃ClN₂O₂
MDL Number: MFCD00490110
CAS Number: 54231-32-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR