6-Nitrobenzothiazole ≥95%

Supplier: Matrix Scientific

072982-5G 072982-10G 072982-100G
102527-766EA 69.23 USD
102527-766 102527-768 102527-770
6-Nitrobenzothiazole ≥95%
6-Nitrobenzothiazole

Hazard: Irritant

Formula: C₇H₄N₂O₂S
MW: 180.19 g/mol
Melting Pt: 173…177 °C
MDL Number: MFCD00014569
CAS Number: 2942-06-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR